Learn More
CTL Scientific Supply Corp QUANT. PERACETIC ACID,100 STR
Supplier: CTL Scientific Supply Corp 91340
QUANTOFIX PERACETIC ACID 50 are test sticks for the semi-quantitative determination of peracetic acid. Gradation 0 - 5 - 10 - 20 - 30 - 50 mg/l. Contains 100 test strips