Learn More
CTL Scientific Supply Corp QUANT ACTIVE OXYGEN,100 TESTS
Supplier: CTL Scientific Supply Corp 91349
QUANTOFIX ACTIVE OXYGEN are test sticks for the semi-quantitative determination of active oxygen. Gradation of scale: 0-4-8-15-25 mg/l PMPS (potassium monopersulphate). Contains 100 test strips