Learn More
MP Biomedicals™ qPCR& Go SYBR No-ROX Kit
Fast and sensitive qPCR SYBR Master Mix. For all other platforms, e.g. Roche, Bio-Rad (not "e;i"e; porfolio), Qiagen, etc.
Supplier:  MP Biomedicals™ 11EBI03050
Catalog No. MP1EBI03050
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • The qPCR& Go SYBR™ No-ROX Kit has been developed for fast, highly reproducible real-time PCR and has been validated on commonly used real-time PCR instruments.
  • A combination of the latest advances in buffer chemistry and enhancers, together with an antibody-mediated hot-start DNA polymerase system, ensures that the qPCR& Go SYBR™ No-ROX Kit delivers fast, highly-specific and ultra-sensitive real-time PCR.
  • For ease-of-use and added convenience, qPCR& Go SYBR™ No-ROX is provided as a 2x mastermix containing all the components necessary for real-time PCR, including the SYBR™ Green I dye, dNTPs, stabilisers and enhancers.
  • The kit consists of a ready-to-use premix, only primers and template need to be added.
Specifications
Specifications
SYBR™green I dye, dNTPs, stabilisers and enhancers. Store at -20°C
qPCR Kit
2x
Fast and sensitive qPCR SYBR Master Mix. For all other platforms, e.g. Roche, Bio-Rad (not “i” porfolio), Qiagen, etc
500 reactions
500
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.