Learn More
Qorpak™ Unlined Round Cans with Triple Tite Lid
Permeation protection against light, oxygen, other gases and essential oils
Supplier:  Qorpak™ MET03084
 View more versions of this product
Catalog No. 02-991-223
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
  • Features resealable multi-friction plug lid for more than one point of contact between lid and can
  • Provides an airtight seal when opened by prying and sealed by pressing
  • May be sterilized by heat
  • Used for paints, varnishes, pastes and adhesives
Specifications
Specifications
Open Top Can
118.2 mL
For paints, varnishes, pastes and adhesives, protection against light, oxygen, other gases and essential oil
Triple Tite Lid
2 in.
2.5 in.
4 oz.
Silver
Metal
520 Cs.
201 mm
Recommendations
Recommendations