Learn More
QBC Diagnostics QBC™ Diagnostics Extended Range Controls
For use with Autoread Plus Hematology System
Supplier: QBC Diagnostics 424305
This item is not returnable.
View return policy
Description
These control samples are designed to meet CLIA requirements for calibration verification. They contain 2 vials each of 3 different blood samples.
Specifications
Autoread Plus Hematology System | |
Extended Range Controls |