Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pymetrozin, Reference Material, MilliporeSigma™ Supelco™

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content incl. uncertainty and expiry date are given on the certificate.

Supplier:  MilliporeSigma Supelco 44184100MG

Catalog No. 11-102-2513


Add to Cart

Description

Description

Specifications

Specifications

123312-89-0
≥98% (HPLC)
100 mg
Safety and Handling

Safety and Handling

H351 - H361fd - H412

ShelfLife : Limited shelf life, expiry date on the label

RTECSNumber : XZ3018620

Recommended Storage : 2°C to 8°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit