Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Puritan™ Puritan Lim Broth Medium
For use in the qualitative procedures for the isolation of Group B Streptococcus (GBS) from clinical specimens as well as in vitro diagnostic use.
$131.00 - $151.00
Specifications
Cap Color | Light Blue |
---|---|
Cap Type | Screw |
Certifications/Compliance | Yes |
Color | Clear Tube |
Content And Storage | 2 to 25°C |
Catalog Number | Mfr. No. | Fill Volume | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Fill Volume | Price | Quantity & Availability | |||||
22-029-649
|
Puritan™
LB200 |
2 mL |
Case of 50 for $131.00
|
|
|||||
22-029-650
|
Puritan™
LB500 |
5 mL |
Case of 50 for $151.00
|
|
|||||
Description
Lim Broth with Colistin and Nalidixic Acid. Screw-cap tube, light blue cap. Selective enrichment medium for use in the qualitative procedures for the isolation of Group B Streptococcus (GBS) from clinical specimens. Selective growth promotion of GBS and inhibition of gram-negative organisms, such as Escherichia coli and Proteus mirabilis. For in Vitro Diagnostic Use. Premium medical grade plastic components to improve product shelf life. For optimum performance, store at 2-25°C.Specifications
Light Blue | |
Yes | |
2 to 25°C | |
United States | |
50 Bulk |
Screw | |
Clear Tube | |
Lim Broth Medium | |
Sterile | |
50 |
Safety and Handling
ShelfLife : 18 Months from DOM
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title