Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Cleaning Applicators

$349.00 - $350.00

Specifications

Length (English) 6 in.
Product Type Applicator
Length (Metric) 15.2cm
Length (English) Handle 6 in.
Length (Metric) Handle 15.2cm
View More Specs

Products 2
Catalog Number Mfr. No. Handle Material Price Quantity & Availability  
Catalog Number Mfr. No. Handle Material Price Quantity & Availability  
19-823-842
View Documents
Puritan™
1806PCFB
Polypropylene
Case for $349.00
Add to Cart
 
19-084795
View Documents
Puritan™
1806WCFL
Wood
Case of 1000 for $350.00
Add to Cart
 
Description

Description

General cleaning swabs. Choose wood or polypropylene handle.
Specifications

Specifications

6 in.
15.2cm
15.2cm
Foam/Cotton
Applicator
6 in.
Non-sterile
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit