Learn More
Cosmo Bio Usa Inc PUREfrex 2.1 | 5 x 250 uL
Supplier:  Cosmo Bio Usa Inc GFKPF2130.255EX
Encompass
PUREfrex (manufactured by Gene Frontier Corporation) is an E. coli reconstituted coupled transcription/translation system based on the PURE system originally developed by the Ueda group (Shimizu et al., 2001) and later commercialized as PURESYSTEM (Shimizu and Ueda, 2010) by BioComber (Tokyo Japan). PUREfrex contains highly purified E. coli proteins comprising modules for aminoacylation, energy regeneration, transcription and translation. Among the many advantages of PUREfrex over competing coupled transcription/translation systems are reduced levels of contaminating proteases, nucleases, and phosphatases, beta-galactosidase and LPS.
Catalog No. NC1604983