Learn More
Tritech Research Inc PUMP-TUBING SET /HI SPEED
SDP
Supplier:  Tritech Research Inc PB4PTSHS
Encompass
Extra Pump-Tubing Set for HIGH SPEED PourBoy 4; The tubing is pharmaceutical grade silicone rubber and can be autoclaved, but the pump itself cannot. TO BE USED ONLY ON PB4 HIGH SPEED UNITS.
Catalog No. NC1666484
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.