Learn More
Fisherbrand™ PTFE Dippers
SDPSCI_ED
Totally inert, reusable.
 View more versions of this product
Catalog No. S37116
Add to Cart
Description
Description
  • Extendable screw-in shafts have a steel core for rigidity
  • Fully encapsulated in PTFE
Specifications
Specifications
Dipper
33.8 oz.
5.21 x 4.04 x 30.2 in. (13.2 x 10.2 x 76.7 cm)
10 cm
76.7 cm
24 in.
61 cm
10.2 cm
PTFE
1000 mL
3.93 in.
30.2 in.
5.21 in.
13.2 cm
4.04 in.
Product Suggestions
Product Suggestions