Learn More
Fisherbrand™ PTFE Crucibles
SDPSCI_ED
Withstand temperatures to 280°C (536°F).
 View more versions of this product
Catalog No. S29203
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
  • Chemically inert
  • Surfaces are machine finished to eliminate contamination and ease cleaning
  • Without covers
Specifications
Specifications
PTFE
72.1 mm
46 mm
Crucible
280°C
100 mL
71 mm
Product Suggestions
Product Suggestions