Learn More
A-M Systems PTFE COAT SILV WIRE 25' .010
Supplier: A-M Systems 787000
This item has a minimum qty of 2 per supplier requirements.
Silver Wire, PTFE; Insulated, Bare Diameter: 0.01 in. 250um, Coated Diameter: 0.013 in. 330um, Nominal AWG: 30(Bare) 28(Coated), feet / Spool: 25, 99.99% Purity and is Offered in a Wide Variety of Sizes for Microelectrode Fabrication