Learn More
A-M Systems PTFE COAT SILV WIRE 25' .008
Supplier: A-M Systems 786500
This item has a minimum qty of 2 per supplier requirements.
Silver Wire, PTFE; Insulated, Bare Diameter: 0.008 in. 200um, Coated Diameter: 0.011 in. 280um, Nominal AWG: 32(Bare) 30(Coated), feet / Spool: 25, 99.99% Purity and is Offered in a Wide Variety of Sizes for Microelectrode Fabrication