Learn More
Labconco™ Protector™ Filtered Fume Hood Accessory, RFID User Authorization Card
For use with Protector Echo and Airo Filtered Fume Hood. Labconco™ Protector™ Filtered Fume Hood Accessory, RFID User Authorization Card are required for use and maintenance providing additional safety and data/usage records.
Supplier: Labconco™ 9580901/EMD
Description
- Radio Frequency Identification User Card
- Required to operate fume hood
- Memory system of hood tracks user fume hood time usage
- Can turn lights and fans on and off
Warranty and Services
One year
Specifications
RFID User Card | |
5.39 cm | |
3.375 in. |
2.125 in. | |
Protector Echo and Airo Filtered Fume Hood | |
8.57 cm |