Learn More
Labconco™ Protector™ Filtered Fume Hood Accessory, gGuard Communication Software
SDPSCI_ED
For use with Protector™ Echo™ and Airo™ Filtered Fume Hood
Supplier:  Labconco™ 9581100/EMD
Catalog No. S28329
Add to Cart
Description
Description
  • Automatically records hood data
  • Alerts and alarms on a customer-supplied personal computer
  • Monitors, manages and provides data via email to designated facility manager
  • Provides access to the following: usage authorization, filter usage, Neutrodine Filter saturation detection, filter identification, sash position, blower speed, laboratory air pollution detection, temperature and usage statistics
  • Software available on CD-ROM
Specifications
Specifications
Software
12.7 cm
0.5 in.
2 in.
5 in.
Protector™ Echo™ and Airo™ Filtered Fume Hood
1.27 cm
5.08 cm