Learn More
Labconco™ Protector™ Filtered Fume Hood Accessory, ADA Remote Control
SDPSCI_ED
For use with Protector Echo™ and Airo™ Filtered Fume Hood. Labconco™ Protector™ Filtered Fume Hood Accessory, ADA Remote Control allows access to fan and light switches through wireless remote control.
Supplier:  Labconco™ 9579700/EMD
Catalog No. S28328
Add to Cart
Description
Description
  • Allows access to fan and light switches through wireless remote control
  • Ideal for persons in wheelchairs and/or shorter individuals
Warranty and Services
One year
Specifications
Specifications
Wireless Remote Control
10.1 cm
1 in.
Lithium battery
5.08 cm
4 in.
Protector™ Echo™ and Airo™ Filtered Fume Hood
2.54 cm
2 in.