Learn More
Propper Self Seal Dust Covers
Supplier: Propper 321212
Description
Satisfies current regulations for sterility maintenance covers.
- Heat sealer not required
- With wide adhesive band for easy sealing procedure
- Linear tear plastic with notches on both sides provides easy opening
- Made up of 3-mil plastic
- Size: 6 x 12 in. (15.24 x 30.48cm)
Specifications
Self Seal Dust Cover | |
Protect the sterile pack during storage, handling, and transportation from dust, moisture or airborne contaminants and extend sterile shelf life when definite shelf life times are used | |
Wide adhesive band, linear tear plastic with notches on both sides |
3 mil Plastic | |
250 | |
Sterilized pack |