Learn More
Propper Autoclave Indicator Tapes
Ideal to secure different types of sterilization packs
Supplier:  Propper 26800407
Catalog No. 11-889-10
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Pressure-sensitive self-adhesive tape
  • Material: crepe-embossed paper
  • Bleading resistant to permanent markers after sterilization
For labeling products for steam and ethylene oxide sterilization
Specifications
Specifications
Autoclavable
Crepe Paper
Steam or Ethyline Oxide sterilization
1 Ea.
54.9 m
Self-adhesive
Autoclave Indicator Tape
Autoclaves
60 yd.
0.5 in.