Learn More
Innovating Science™ Properties of Toothpaste
Investigate several properties of toothpaste samples.
Supplier: Innovating Science™ IS9350
Alert:
WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, which is known to the State of California to cause cancer.
Description
- Investigate the presence of fluoride, pH, abrasiveness and foaming ability
Order Info
Class size: 15 groups.
Specifications
49 CFR 173.4 | |
White | |
13 to 20 | |
1 x 30mL Hydrochloric Acid, 1 x 100mL Glycerin, 1 x 25g Sodium Lauryl Sulfate, 1 x 25g gum, arabic, 1 x 150g Calcium Carbonate, 3 tubes toothpaste, 1 universal indicator strips, 1 Fluoride test strips, 1 Acetate sheets, 45 Polystyrene test tubes, 45 gradu | |
12 x 9.25 x 4 in. |
Properties of Toothpaste Kit | |
7 to 14 | |
To investigate several properties of toothpaste samples | |
1 | |
15 Lab Groups |