Learn More
Master Appliance PROHEAT VARITEMP PH-1200PROHE
Supplier:  Master Appliance 10207
Encompass_Preferred
Proheat Varitemp Heat Gun, 126degreeF (52degreeC) to 803degreeF (428degreeC) +/- 10%, 120V, 60Hz, 12A Max., 1500w Max., Compact, lightweight, easy-to-use variable temperature heat gun, Ergonomic design for superb balance and grip
Catalog No. NC9036443
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.