Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone, Wettable, Powder, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog Number 18605854
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
57-83-0
C21H30O2
314.47
RJKFOVLPORLFTN-LEKSSAKUSA-N
(1S,3aS,3bS,9aR,9bS,11aS)-1-acetyl-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
57-83-0
C21H30O2
314.47
RJKFOVLPORLFTN-LEKSSAKUSA-N
(1S,3aS,3bS,9aR,9bS,11aS)-1-acetyl-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

PR111, 57-83-0, C21H30O2

Spectrum™ Chemical Progesterone, Wettable, Powder, USP
CAS Max % 1
Loss on Drying 0.005
Melting Range 126-131°C
Packaging Amber Glass Bottle
Quantity 1 kg
Specific Rotation +175° to +183°
Percent Purity 97 to 103%
Grade USP
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit