Learn More
MESA LABS BOZEMAN MFG FACILITY Class 5 Moving Front Integrator, 250/Pack
Supplier: MESA LABS BOZEMAN MFG FACILITY 7705CISSW
PROCHEM SSW – STEAM TYPE 5 INTEGRATING INDICATOR. Moving front Integrator, no interpretation necessary. Effective across the normal sterilization range. Parallels the performance of a biological indicator. Conforms to applicable portions of ISO 11140-1. 250 indicators/box. 5 year expiry.