Learn More
Pendotech Pendotech Pressuremat Monitor, Transmitter, And Alarm (Pmat2F)
Supplier: Pendotech PMAT2F
PressureMAT PLUS monitor, transmitter, alarm for two single use pressure sensor input and two flow meter input (4 analog outputs)