Learn More
Pendotech Pendotech Pressuremat Monitor, Transmitter, And Alarm (Pmat-S)
Supplier: Pendotech PMATS
PressureMAT monitor, transmitter, alarm for 1 single use pressure sensor (analog 4-20 mA output, alarm relay output, tare input)