Learn More
Pendotech Pendotech Pressuremat Monitor, Transmitter, And Alarm (Pmat3A)
Supplier:  Pendotech PMAT3A
Encompass_Preferred
PressureMAT PLUS monitor, transmitter, alarm for three single use pressure sensor inputs and one 4-20mA analog input (4 analog outputs)
Catalog No. NC0764368