Learn More
SWAGELOK CAMBRIDGE PRESSURE GAUGE
Supplier:  SWAGELOK CAMBRIDGE PGI63BPG300LAOXJ
Encompass
Industrial Pressure Gauge, Adj Pointer, 63 mm, 0 to 300 psi, bar secondary, Lower Mount, 1/4 in. MNPT
Catalog No. NC1473421