Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B Medical Systems™ Premium Pharmacy Freezer, 949 L (33.5 ft.3)
Our Premium pharmacy freezers offer intuitive touch-screen controls and remote alarm monitoring for reliable -41 to -20°C storage.
Supplier: B Medical Systems™ F900
Description
Premium and Precision Series laboratory and pharmacy freezers are medical-grade freezers designed to preserve temperature-sensitive enzymes, media, and clinical specimens.
- Freezers include positive airflow rear wall plenums and interior designs, allowing ample clearance around inventory loads to assure uniform storage temperatures and overall stability
- Advanced refrigeration systems include reserve refrigeration power for operation in high ambient installations to maintain temperature stability, uniformity, and fast recovery
Specifications
Lab Freezer | |
949 L | |
Solid | |
Polyurethane Foam | |
Painted Steel | |
Upright | |
Air Cooled | |
Lab and Pharmacy Storage | |
Manual | |
40.9 x 44.8 x 78.3 in. | |
40.9 in. | |
78.3 in. | |
B Connected | |
CO2 or LN2 Optional | |
NEMA 5-20 | |
60 Hz | |
1300 w | |
Bottom-mounted | |
776 lb. |
33.5 cu. ft. | |
-41°C to -20°C | |
1 (2 compartments) | |
Stainless Steel | |
cUL, FDA Class I, ENERGYSTAR | |
Off-white | |
Touch Screen | |
7 Year Compressor, 2 Year Parts and Labor | |
F900 | |
738 x 887 x 1173 mm (29.05 x 34.92 x 46.18 in.) | |
29 in. | |
44.8 in. | |
Alarm system | |
20 A | |
115 V | |
1 | |
7.6 kWh/day | |
R290 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction