Learn More
MCR Safety Predator™ PD2 Safety Glasses
Click to view available options
Lens Tint:
Clear
Gray
Description
- Meets the ANSI/ISEA Z87.1 D4 designation for dust protection and ANSI/ISEA Z87.1 for high impact
- Full foam venting helps to reduce heat buildup and assists in minimizing fogging
- Available in standard anti-fog or MAX6™ superior anti-fog
- Wide profile lens provides excellent peripheral vision
- Elastic strap with hook and loop, and 5-point ratchet provide complete adjustability
Specifications
Description | Dialectric, Direct Vented, Perforated |
Lens Tint | Clear |
Frame Material | Polyurethane |