Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide Solution, 30%, Fisher Chemical

Catalog Number FL20135
Click to view available options
Quantity:
5 gal.
1310-58-3
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
14797
CHEBI:32035
potassium hydroxide
[OH-].[K+]
1310-58-3
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
14797
CHEBI:32035
potassium hydroxide
[OH-].[K+]
pH 12.0
Color Colorless
Melting Point -28.8°C
Density 1.510g/cm³
Quantity 5 gal.
Percent Purity 29.5 to 30.5%
Identification Pass Test
Vapor Density >1.0g/mol
Vapor Pressure 2.6mmHg at 20°C
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide Solution, 30%
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit