Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Hydroxide, DILUT-IT™ Analytical Concentrate, 1N, 1 equiv. = 56.11g KOH, J.T. Baker™
Click to view available options
Quantity:
1 Pk.
Packaging:
Poly Ampoule
Specifications
Quantity | 1 Pk. |
Packaging | Poly Ampoule |
Specific Gravity | 1L = 1.05kg |
Chemical Name or Material | Potassium Hydroxide, DILUT-IT™ Analytical Concentrate, 1N, 1 equiv. = 56.11g KOH |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction