Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.1 N Alcoholic Solution in Methanol, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
1310-58-3, 67-56-1
1310-58-3, 67-56-1
Concentration 0.1 N ± 0.005 N
Quantity 20 L
Grade Chemical Solution
Packaging Cubitainer

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.