Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Hydrogen Phthalate, Acidimetric Standard, Crystal, Reagent Special, ACS, 99.95-105%, Spectrum™ Chemical
Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Potassium Hydrogen Phthalate, Acidimetric Standard, Crystal, Reagent Special, ACS is used in analytical chemistry as a disintegrating agent can be used as a buffering agent (in combination with hydrochloric acid (HCl) or sodium hydroxide (NaOH). As an ACS grade Reagent, Spectrum Chemical manufactured Potassium Hydrogen Sulfate is used as the quality standard against which other Potassium Hydrogen Sulfate substances are graded. it has met the toughest regulatory standards for quality and pureness.
Specifications
CAS Max % | 1 |
pH | 4.00 to 4.02 |
Packaging | Fiber Drum |
Quantity | 50 kg |
Percent Purity | 99.95 to 100.05% |
Grade | ACS |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction