Learn More
Aqua Solutions POTASSIUM CHLORIDE 10UM 500ML
Supplier: Aqua Solutions 6921500ML
Conductivity Standard Traceable to N.I.S.T, 500 mL, Component: Potassium Chloride, 7447-40-7, <1%, Water, Deionized ASTM Type II, 7732-18-5, Balance, MP: ~0degreeC, BP: ~100degreeC, Specific Gravity: 1, Volatile by Volume: > 99%, Appearance