Learn More
CTL Scientific Supply Corp POT IODIDE STAR PAP,PK/3 DISP
Supplier: CTL Scientific Supply Corp 907543
POTASSIUM IODIDE STARCH PAPER - Pack of 3 dispensers each containing a 5 meter roll of paper This is a filter paper impregnated with potassium iodide (KI) and starch for the detection of nitrite & free chlorine. Nitrite or free chlorine oxidize potassium iodide to form elemental iodine which reacts with the starch in known manner.