Learn More
Thermo Scientific Pot.Data Uncoded Combo Lamp
Supplier: Thermo Scientific 942339321021
Na/K Uncoded. For Use With (Equipment) iCE 3000 Series Atomic Absorption Spectrometers. Type Hollow Cathode Lamps, Uncoded