Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ 50 HQ RoboColumn™ Pre-packed columns
SDP

Thermo Scientific™ RoboColumn™ chromatography products are small columns provided in 8-column strips.

$262.00 - $341.00

Specifications

Column Type Anion Exchange Column
Content And Storage Store at 2°C to 8°C or at room temperature.
For Use With (Application) Anion Exchange, Protein Purification
Max. Pressure 116 psi (8 bar)
View More Specs

Products 2
Catalog Number Mfr. No. Column Capacity Price Quantity & Availability  
Catalog Number Mfr. No. Column Capacity Price Quantity & Availability  
A30717
SDP
View Documents
Thermo Scientific™
A30717
200 μL
Each for $262.00
Add to Cart
 
A30718
SDP
View Documents
Thermo Scientific™
A30718
600 μL
Each for $341.00
Add to Cart
 
Description

Description

RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS HQ50 resin, a 50-micron strong anion-exchange resin suitable for low-pressure applications, including large-scale downstream bioprocessing.

POROS HQ is a high-performance chromatography resin based on a quaternized polyethyleneimine functional group, yielding a high capacity. It is designed for the separation and purification of biomolecules for analytical to process scale separations

Features of the resin:

  • Higher productivity—through high throughput and high dynamic capacity associated with high performance chromatography
  • Chemical stability—allows aggressive cleaning and sanitization
  • Enhanced biomolecule access—provided via large pores (500-10,000 Å)
  • Polystyrenedivinylbenzene particles—yield a robust, easily packable matrix
  • Better separation methods in less time—the speed of high performance chromatography reduces weeks of experimentation to only a few hours, leaving more time to explore all the variables of your separation

RoboColumn™ is a registered trademark of ATOLL GmbH.

Specifications

Specifications

Anion Exchange Column
Store at 2°C to 8°C or at room temperature.
Anion Exchange, Protein Purification
116 psi (8 bar)
SDS
Documents

Documents

Product Certifications
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.