Learn More
STELLEN MEDICAL LLC Porcine Skin Sheet 1-24 sheets Size: 8" x 8" (20 cm x 20 cm)
Supplier: STELLEN MEDICAL LLC I188124 SHEETS
porcine skin sheet 1-24 sheets Size: 8" x 8" (20 cm x 20 cm) Individually wrapped, shaved and frozen, non-sterile skin • Consistent thickness 0.060” +/- 0.010” (1.524 mm +/- 0.254 mm) thick • Split thickness (epidermal and dermal surfaces; epidermal side faces product labeling) • Includes notch to indication orientation of skin • Frozen item shipped on dry ice for overnight delivery via FedEx Store in a standard freezer (less than 14F to -13F) Porcine Skin, Non-Sterile, Store Frozen (USED FOR TESTING PURPOSES)NOT FOR HUMAN USE
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.