Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Applied Biosystems™ POP-4™ Polymer for the 310 Genetic Analyzer
POP-4™is a denaturing polymer used for the 310 Genetic Analyzer.
Manufacturer: Applied Biosystems™ 402838

Includes: 1mL Glass Syringe, 310 Genetic Analyzer Capillary, 47cm (36cm wtr), 310 Genetic Analyzer Capillary, 61cm (50cm wtr), 310 Running Buffer, 10X, AmpFLSTR™ Yfiler™ PCR Amplification Kit, AmpFLSTR™ Identifiler™ PCR Amplification Kit, AmpFLSTR™ SGM Plus™ PCR Amplification Kit, AmpliTaq Gold™ DNA Polymerase, 1000 units with Buffer II and MgCl2 solution, BigDye™ Terminator v1.1 Cycle Sequencing Kit, GeneScan™ 400HD ROX™ Size Standard, GeneScan™ 500 LIZ™ Size Standard, GeneScan™ 500 ROX™ Size Standard, Genetic Analyzer Sample Tubes, 0.5mL, Hi-Di™ Formamide, MicroAmp™ 8-Tube Strip, 0.2mL, MicroAmp™ Reaction Tube with Cap, 0.2mL, POP-4™ Polymer for 3130/3130xl Genetic Analyzers, POP-6™ Polymer for the 310 Genetic Analyzer, Quantifiler™ Human DNA Quantification Kit, Septa for 0.5mL Sample Tubes
Description
- Easy to Use: Pre-formulated, which not only saves time but also saves reproducibility by ensuring consistent formulation
- Robust Formulation: Formulated for sequencing and fragment analysis applications
- Polymers allow capillaries to be used multiple times
Specifications
One bottle. Store at 2°C to 8°C. | |
Fragment Analysis (Denaturing), Resequencing | |
SD <0.15 nt to 250 nt (for 36 cm capillary) | |
DNA Sequencing Reagents | |
1 base detection up to 250 nt |
310 Genetic Analyzer | |
Stability on instrument 7 days, Up to 500 samples/5 ml | |
POP-4™ | |
5 mL |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For research use only