Learn More
Polywax™ 655, Analytical Standard, MilliporeSigma™ Supelco™
Polywax™ 655, a polyethylene wax, is a high molecular weight hydrocarbon standard for high temperature simulated distillation or gas chromatography analysis.
Supplier: MilliporeSigma Supelco 48477
Specifications
Neat | |
Analytical Standard | |
5000 mg |
Polywax(R) | |
Ampule |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Polywax is a registered trademark of Baker Hughes, Inc