Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polymedco™ Sed-Chek™ 2 ESR Controls for StaRRsed Analyzers (Select Models)
Whole blood reference control materials designed to monitor ESR procedures
$274.00
Specifications
Cap Type | Automatic self-zeroing |
---|---|
Compatible Methods | Both automated and manual ESR methods |
Content And Storage | 20°C to 22°C |
For Use With (Equipment) | StaRRsed analyzer |
Intended Use | Whole Blood reference control material designed to monitor ESR procedures |
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
21-000-120
|
Polymedco™
ESR2SSA |
Abnormal Control |
Pack of 6 for $274.00
|
|
|||||
21-000-121
|
Polymedco™
ESR2SSN |
Normal Control |
Pack of 6 for $274.00
|
|
|||||
Description
Controls help to monitor technique as well as environmental, physical, and mechanical factors such as room temperature, and tube position, vibration, or instrument problems. With an advanced formula, these controls are comprehensive and assayed for both automated and manual methods.
- 18 month closed-vial stability from date of manufacture
- 31 day open-vial stability
- Easy-to-mix formula
- Available in 6 x 5mL vials
Specifications
Automatic self-zeroing | |
20°C to 22°C | |
Whole Blood reference control material designed to monitor ESR procedures | |
6 x 5 mL |
Both automated and manual ESR methods | |
StaRRsed analyzer | |
Vial | |
31 Days Open Vial |
Safety and Handling
ShelfLife : 18 Months
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title