Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polyethyleneglycol 60 Castor Oil, Hydrogenated, Spectrum™ Chemical
Click to view available options
Quantity:
500 g
18 kg
450 lb.
Packaging:
Amber Glass Bottle
Drum
Description
Spectrum™ Chemical Polyethyleneglycol 60 Castor Oil, Hydrogenated is a polyether compound that is used in a wide variety of fields including pharmaceutical manufacturing as an excipient and active ingredient. Due to its low toxicity it can be used as a lubricating coating for various surfaces in aqueous and non-aqueous environments, a reagent in biochemistry to create very high osmotic pressures, a polar stationary phase for gas chromatography and as a binder. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Color | Off-white solid |
CAS Max % | 1 |
Acid Value | 1.5 |
Quantity | 500 g |
Grade | Reagent |
Hydroxyl Value | 50-70 |
Packaging | Amber Glass Bottle |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction