Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Polyethylene, Tap (Attachable)
Click to view available options
Polyethylene
Specifications
Product Type | Tap |
Closure Size | 60x6 Thread |
Description | Self-venting, attachable tap |
Material | Polyethylene |
For Use With (Equipment) | 5 L, 10 L and 25 L PE Cannisters |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction