Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Hemoglobin Reagents
$53.75 - $100.00
Specifications
Certifications/Compliance | CE Marked |
---|---|
Type | Hemoglobin Reagent Set |
Formulation | Total hemoglobin reagent: Potassium Ferricyanide 0.6mM, Potassium Cyanide 0.77mM, Buffers and Stabilizers |
For Use With (Application) | Quantitative determination of Hemoglobin in whole Blood |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-666-305
|
Pointe Scientific
H7504120 |
1 x 120 mL |
Each for $53.75
|
|
|||||
23-666-306
|
Pointe Scientific
H7504500 |
1 x 500 mL |
Each for $100.00
|
|
|||||
Specifications
CE Marked | |
Total hemoglobin reagent: Potassium Ferricyanide 0.6mM, Potassium Cyanide 0.77mM, Buffers and Stabilizers | |
Liquid |
Hemoglobin Reagent Set | |
Quantitative determination of Hemoglobin in whole Blood |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title