Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific γ-Glutamyl Transferase Liquid Reagents
$205.00 - $554.00
Specifications
Certifications/Compliance | CE Marked |
---|---|
Type | gamma-Glutamyl Transferase Reagent Kit |
Formulation | Tris Buffer 89mM/L pH 8.1, Glycylglycine 126mM/L, GLUPA-C 3.3mM/L, Sodium Azide 0.095% |
For Use With (Application) | Quantitative kinetic determination of gamma Glutamyl Transferase activity in Serum |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-666-256
|
Pointe Scientific
G7571150 |
1 x 120mL R1, 1 x 30mL R2 |
Each for $205.00
|
|
|||||
23-666-258
|
Pointe Scientific
G7571625 |
1 x 500mL R1, 1 x 125mL R2 |
Each for $554.00
|
|
|||||
23-666-257
|
Pointe Scientific
G7571450 |
3 x 120mL R1, 1 x 90mL R2 |
Each for $451.00
|
|
|||||
Specifications
CE Marked | |
Tris Buffer 89mM/L pH 8.1, Glycylglycine 126mM/L, GLUPA-C 3.3mM/L, Sodium Azide 0.095% | |
Liquid |
gamma-Glutamyl Transferase Reagent Kit | |
Quantitative kinetic determination of gamma Glutamyl Transferase activity in Serum |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title