Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Creatinine (Cold Stable) Reagents
$56.50 - $182.00
Specifications
Certifications/Compliance | CE Marked |
---|---|
Type | Creatinine Reagent Set |
Formulation | Creatinine R1: Alkaline Buffer, Creatinine R2: Picric Acid 40mM, Surfactant |
For Use With (Application) | Quantitative determination of Creatinine in Serum, in-vitro diagnostic use only |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-666-236
|
Pointe Scientific
C75391250 |
1 x 1L R1, 1 x 250mL R2 |
Each for $182.00
|
|
|||||
23-666-237
|
Pointe Scientific
C7539150 |
1 x 120mL R1, 1 x 30mL R2 |
Each for $56.50
|
|
|||||
23-666-238
|
Pointe Scientific
C7539625 |
1 x 500mL R1, 1 x 125mL R2 |
Each for $127.00
|
|
|||||
Description
- Reagent requires Creatinine Standard (23-666-235)/Chemistry Calibrator (23-666-189)
Specifications
CE Marked | |
Creatinine R1: Alkaline Buffer, Creatinine R2: Picric Acid 40mM, Surfactant | |
Liquid |
Creatinine Reagent Set | |
Quantitative determination of Creatinine in Serum, in-vitro diagnostic use only |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title