Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Calcium (Arsenazo III) Reagents
Quantify calcium levels in serum or heparinized plasma
$53.25 - $267.00
Specifications
Certifications/Compliance | CE Marked |
---|---|
Type | Calcium Arsenazo III Reagent Kit |
Formulation | Calcium reagent: Arsenazo III <0.15mM, 8-Hydroxyquinoline Sulfonate 5mM, Buffer, Surfactant |
For Use With (Application) | Quantitative determination of Calcium in Serum or Heparinized Plasma |
Wavelength | 650 nm |
Includes: Calcium reagent: Arsenazo III≥0.15mM, 8-Hydroxyquinoline Sulfonate 5.0mM, buffer, surfactant
Requires: Requires Calibrator/Standard
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-666-170
|
Pointe Scientific
C7529120 |
1 x 120 mL |
Each for $53.25
|
|
|||||
23-666-172
|
Pointe Scientific
C7529500 |
1 x 500 mL |
Each for $164.00
|
|
|||||
23-666-171
|
Pointe Scientific
C75291L |
1 x 1 L |
Each for $267.00
|
|
|||||
Description
- Method: Arsenazo III, Endpoint
- Format: Liquid (ready to use)
- Wavelength: 650nm
- Linearity: 0 to 15mg/dL
- Expected Values: 8.5 to 10.4mg/dL
For the quantitative determination of Calcium in serum or heparinized plasma.
Specifications
CE Marked | |
Calcium reagent: Arsenazo III <0.15mM, 8-Hydroxyquinoline Sulfonate 5mM, Buffer, Surfactant | |
650 nm |
Calcium Arsenazo III Reagent Kit | |
Quantitative determination of Calcium in Serum or Heparinized Plasma | |
Liquid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title