Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Alcohol Reagents
$182.00 - $289.00
Specifications
Certifications/Compliance | CE Marked |
---|---|
Type | Alcohol Reagent Kit |
Formulation | Alcohol Dehydrogenase (Yeast) 200000U/L, NAD 2.6mM, Buffer pH 9.2 ± 0.1, Surfactant, preservative |
For Use With (Application) | Quantitative determination of Ethyl Alcohol in Serum or whole Blood, in-vitro diagnostic use only |
Form | Dry Powder |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-666-073
|
Pointe Scientific
A750439 |
6 x 6.5 mL |
Pack of 6 for $182.00
|
|
|||||
23-666-072
|
Pointe Scientific
A7504150 |
10 x 15 mL |
Pack of 10 for $289.00
|
|
|||||
Specifications
CE Marked | |
Alcohol Dehydrogenase (Yeast) 200000U/L, NAD 2.6mM, Buffer pH 9.2 ± 0.1, Surfactant, preservative | |
Dry Powder |
Alcohol Reagent Kit | |
Quantitative determination of Ethyl Alcohol in Serum or whole Blood, in-vitro diagnostic use only |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title