Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podophyllum Resin, USP, 40-50%, Spectrum™ Chemical
SDP

Catalog Number 18605256
Click to view available options
Quantity:
5 g
25 g
125 g
1 kg
Packaging:
Amber Glass Bottle
9000-55-9
C88H92O34
1693.67
KOUZYZCESNTXJO-UHFFFAOYNA-N
bis(16-hydroxy-10-(3,4,5-trimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one); bis(8-hydroxy-7-(hydroxymethyl)-5-(3,4,5-trimethoxyphenyl)-2H,5H,6H,7H,8H-naphtho[2,3-d][1,3]dioxole-6-carboxylic acid)
COC1=CC(=CC(OC)=C1OC)C1C2C(COC2=O)C(O)C2=CC3=C(OCO3)C=C12.COC1=CC(=CC(OC)=C1OC)C1C2C(COC2=O)C(O)C2=CC3=C(OCO3)C=C12.COC1=CC(=CC(OC)=C1OC)C1C(C(CO)C(O)C2=CC3=C(OCO3)C=C12)C(O)=O.COC1=CC(=CC(OC)=C1OC)C1C(C(CO)C(O)C2=CC3=C(OCO3)C=C12)C(O)=O
9000-55-9
C88H92O34
1693.67
KOUZYZCESNTXJO-UHFFFAOYNA-N
bis(16-hydroxy-10-(3,4,5-trimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one); bis(8-hydroxy-7-(hydroxymethyl)-5-(3,4,5-trimethoxyphenyl)-2H,5H,6H,7H,8H-naphtho[2,3-d][1,3]dioxole-6-carboxylic acid)
COC1=CC(=CC(OC)=C1OC)C1C2C(COC2=O)C(O)C2=CC3=C(OCO3)C=C12.COC1=CC(=CC(OC)=C1OC)C1C2C(COC2=O)C(O)C2=CC3=C(OCO3)C=C12.COC1=CC(=CC(OC)=C1OC)C1C(C(CO)C(O)C2=CC3=C(OCO3)C=C12)C(O)=O.COC1=CC(=CC(OC)=C1OC)C1C(C(CO)C(O)C2=CC3=C(OCO3)C=C12)C(O)=O

PO105, 9000-55-9

Spectrum™ Chemical Podophyllum Resin, USP is used as a medication to treat genital warts and plantar warts. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Ignition Residue 0.015
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 40 to 50%
Grade USP
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit