Learn More
Pluriselect pluriStrainer Membrane-Mini (Cell Strainer) 3 um pore-size, 5 pcs. , sterile
Supplier: Pluriselect 461000305
Membrane filters have a well-defined pore size exclusion limit (available with 1, 3, 5 and 8 um pores), allowing for precise filtration and detection of very small particles. The membranes are non-hygroscopic (optimal for sample recovery), have low non-specific protein binding (reduces sample loss in protein-containing samples), are biologically inert, non-cytotoxic, and have excellent chemical and temperature resistance. The membrane filters can be used for example in biological or gravimetric analysis, cytology, parasitology or in environmental analyzes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.